Furil 98%
Supplier: Thermo Scientific Chemicals
A15386-06
A15386-14
CAAAA15386-06EA
164.32
CAD
CAAAA15386-06
CAAAA15386-14
CAAAA15386-22
Furil 98%
Furil
Purity: 98%
CAS No.: 492-94-4
Molecular Formula: C10H6O4
Formula Weight: 190.15
Melting Point: 162-165°
Beilstein Reference: 383882
MDL No.: MFCD00003241
Formula:
C₁₀H₆O₄ Melting Pt: 162…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003241 CAS Number: 492-94-4 EINECS: 207-766-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...