4-tert-Butylbenzamide 98%

Supplier: Thermo Scientific Chemicals
Warning

A15380-09 A15380-18
CAAAA15380-09EA 120.4 CAD
CAAAA15380-09 CAAAA15380-18 CAAAA15380-30
4-tert-Butylbenzamide 98%
4-tert-Butylbenzamide

Purity: 98% CAS No.: 56108-12-4 Molecular Formula: C11H15NO Formula Weight: 177.25 Melting Point: 171-175° Beilstein Reference: 2086692

Formula: C₁₁H₁₅NO
Melting Pt: 171…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00017139
CAS Number: 56108-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR