Copper(II) oxalate hemihydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cupric oxalate hemihydrate

A15365-18 A15365-30
CAAAA15365-18EA 103.65 CAD
CAAAA15365-18 CAAAA15365-30
Copper(II) oxalate hemihydrate 98%
Copper(II) oxalate hemihydrate

Soluble in NH₃OH


Purity: 98% CAS No.: 5893-66-3 Molecular Formula: CuC2O4.0.5H2O Formula Weight: 160.57 (151.51anhy) Solubility: Soluble in NH4OH UN No.: 3288 MDL No.: MFCD00150451

Formula: CuC₂O₄.½H₂O
MW: 160.57 g/mol (151.51 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150451
CAS Number: 5893-66-3
EINECS: 212-411-4
UN: 3288
ADR: "6.1",III
Merck Index: 14,2645

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR