6-Bromo-2-oxo-2H-chromene-3-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Bromocoumarin-3-carboxylic acid, UBP608
A15354-06
A15354-14
CAAAA15354-06EA
154.52
CAD
CAAAA15354-06
CAAAA15354-14
CAAAA15354-22
6-Bromo-2-oxo-2H-chromene-3-carboxylic acid 97%
6-Bromo-2-oxo-2H-chromene-3-carboxylic acid
Purity: 97%
CAS No.: 2199-87-3
Molecular Formula: C10H5BrO4
Formula Weight: 269.05
Melting Point: 194-196°
Beilstein Reference: 13960
MDL No.: MFCD00047640
Formula:
C₁₀H₅BrO₄ MW: 269.06 g/mol Melting Pt: 194…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047640 CAS Number: 2199-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...