Methyl-3-amino-4-methylbenzoate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-amino-4-methylbenzoate

A15348-06 A15348-14 A15348-22
CAAAA15348-06EA 44.87 CAD
CAAAA15348-06 CAAAA15348-14 CAAAA15348-22
Methyl-3-amino-4-methylbenzoate 97%
Methyl 3-amino-4-methylbenzoate

Purity: 97% CAS No.: 18595-18-1 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Melting Point: 113-117° Beilstein Reference: 2088611

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00025206
CAS Number: 18595-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR