Dicyclohexano-18-crown-6 (mixture of isomers) 97%

Supplier: Thermo Scientific Chemicals
Danger

A15344-03 A15344-09 A15344-14
CAAAAA15344-03EA 50.88 CAD
CAAAAA15344-03 CAAAAA15344-09 CAAAAA15344-14
Dicyclohexano-18-crown-6 (mixture of isomers) 97%
Dicyclohexano-18-crown-6 (mixture of isomers)

Fieser: 4,142 5,207 6,174 9,126


Purity: 97% CAS No.: 16069-36-6 Molecular Formula: C20H36O6 Formula Weight: 372.50 Flash Point: 110°(230°F) Odor: Odorless Beilstein Reference: 1130529 Merck Reference: 14,2602 UN No.: 2811 MDL No.: MFCD00005099

Formula: C₂₀H₃₆O₆
MW: 372.51 g/mol
Melting Pt: 46…53 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005099
CAS Number: 16069-36-6
EINECS: 240-216-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,2602

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR