Alloxan monohydrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15324-14
A15324-22
A15324-18
A15324-30
CAAAAA15324-14EA
86.43
CAD
CAAAAA15324-14
CAAAAA15324-22
CAAAA15324-0B
CAAAA15324-18
CAAAA15324-30
Alloxan monohydrate 98%
Alloxan monohydrate
Cytotoxic compound that causes oxidative base damage to nuclear DNA
Purity: 98%
CAS No.: 2244-11-3
Molecular Formula: C4H2N2O4.H2O
Formula Weight: 160.09 (142.07anhy)
Storage Temperature: 2°C to 8°C
Melting Point: ca 260° dec.
Beilstein Reference: 5309394
MDL No.: MFCD00149399
Formula:
C₄H₂N₂O₄ Melting Pt: ca 260 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00149399 CAS Number: 2244-11-3 EINECS: 200-062-0 Merck Index: 14,282 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...