Ethylenediaminetetraacetic acid diammonium salt hydrate

Supplier: Thermo Scientific Chemicals

A15286-14 A15286-22 A15286-36
CAAAA15286-14EA 55.3 CAD
CAAAA15286-14 CAAAA15286-22 CAAAA15286-36
Ethylenediaminetetraacetic acid diammonium salt hydrate
Ethylenediaminetetraacetic acid diammonium salt hydrate

CAS No.: 20824-56-0 Molecular Formula: (NH4O2CCH2)2NCH2CH2N(CH2CO2H)2.xH2O Formula Weight: 326.31(anhy) Beilstein Reference: 7785165 MDL No.: MFCD00150463

Formula: (NH₄O₂CCH₂)₂NCH₂CH₂N(CH₂CO₂H)₂.xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00150463
CAS Number: 20824-56-0
EINECS: 244-063-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR