3-Cyclohexene-1-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cyclohex-3-enecarboxylic acid, 3-Cyclohexenecarboxylic acid, 1,2,3,6-Tetrahydrobenzoic Acid

A15229-06 A15229-14
CAAAAA15229-06EA 129.56 CAD
CAAAAA15229-06 CAAAAA15229-14
3-Cyclohexene-1-carboxylic acid 98%
3-Cyclohexene-1-carboxylic acid

V sol


Purity: 98% CAS No.: 4771-80-6 Molecular Formula: C7H10O2 Formula Weight: 126.16 Flash Point: 110°(230°F) Refractive Index: 1.4815 Melting Point: 16-18° Boiling Point: 120-122°/10mm Solubility: V sol Beilstein Reference: 1617723 UN No.: 3265 MDL No.: MFCD00013781

Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Boiling Pt: 120…122 °C/10 mm
Melting Pt: 16…18 °C
Density: 1.082
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013781
CAS Number: 4771-80-6
EINECS: 225-314-7
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR