Thiophene-2-carboxamide 99%

Supplier: Thermo Scientific Chemicals
Warning

A15219-06 A15219-14
CAAAA15219-06EA 66.04 CAD
CAAAA15219-06 CAAAA15219-14
Thiophene-2-carboxamide 99%
Thiophene-2-carboxamide

Purity: 99% CAS No.: 5813-89-8 Molecular Formula: C5H5NOS Formula Weight: 127.17 Melting Point: 178-183° Beilstein Reference: 110153

Formula: C₅H₅NOS
Melting Pt: 178…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00010425
CAS Number: 5813-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR