Pamoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A15207-18 A15207-30
CAAAA15207-18EA 95.12 CAD
CAAAA15207-18 CAAAA15207-30
Pamoic acid 99%
Pamoic acid

Used in preparation of repository derivatives of medicinal agents


Purity: 99% CAS No.: 130-85-8 Molecular Formula: C23H16O6 Formula Weight: 388.38 Melting Point: 300° Solubility: Soluble in nitrobenzene, pyridine Beilstein Reference: 901319 MDL No.: MFCD00004079


Applications: Used in preparation of repository derivatives of medicinal agents

Formula: C₂₃H₁₆O₆
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004079
CAS Number: 130-85-8
EINECS: 204-998-0
Merck Index: 14,7005

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR