Diethyl (4-methylbenzyl)phosphonate 98+%

Supplier: Thermo Scientific Chemicals

A15190-09 A15190-18
CAAAA15190-09EA 54.83 CAD
CAAAA15190-09 CAAAA15190-18 CAAAA15190-30
Diethyl (4-methylbenzyl)phosphonate 98+%
Diethyl (4-methylbenzyl)phosphonate

Purity: 98+% CAS No.: 3762-25-2 Molecular Formula: C12H19O3P Formula Weight: 242.26 Flash Point: 110°(230°F) Refractive Index: 1.4960 Boiling Point: 146-148°/0.6mm Beilstein Reference: 2940626 MDL No.: MFCD00041385

Formula: C₁₂H₁₉O₃P
Boiling Pt: 146…148 °C/0.6 mm
Density: 1,070
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041385
CAS Number: 3762-25-2
EINECS: 223-180-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR