Ethyl trimethylacetopyruvate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15164-03
A15164-06
CAAAAA15164-14EA
3556.07
CAD
CAAAAA15164-14
CAAAA15164-03
CAAAA15164-06
Ethyl trimethylacetopyruvate 98%
Ethyl trimethylacetopyruvate
Purity: 98%
CAS No.: 13395-36-3
Molecular Formula: C10H16O4
Formula Weight: 200.24
Refractive Index: 1.4700
Boiling Point: 68-70°/0.5mm
MDL No.: MFCD00052319
Formula:
C₁₀H₁₆O₄ Boiling Pt: 68…70 °C/0.5 mm Storage Temperature: Ambient |
MDL Number:
MFCD00052319 CAS Number: 13395-36-3 EINECS: 236-478-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...