N,N-Dimethylmethyleneammonium iodide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15146-09
A15146-18
A15146-30
CAAAA15146-09EA
156.1
CAD
CAAAA15146-09
CAAAA15146-18
CAAAA15146-30
N,N-Dimethylmethyleneammonium iodide 97%
N,N-Dimethylmethyleneammonium iodide
Fieser: 4,186 8,194 10,160
Purity: 97%
CAS No.: 33797-51-2
Molecular Formula: C3H8IN
Formula Weight: 185.01
Storage Temperature: 2°C to 8°C
Melting Point: ca 240° dec.
Beilstein Reference: 3594966
MDL No.: MFCD00011810
Formula:
C₃H₈IN Melting Pt: ca 240 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00011810 CAS Number: 33797-51-2 EINECS: 251-680-2 Merck Index: 14,3251 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...