1,5-Diphenyl-2,4-pentadien-1-one 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cinnamylideneacetophenone

A15114-06 A15114-14
CAAAA15114-06EA 35.23 CAD
CAAAA15114-06 CAAAA15114-14 CAAAA15114-22
1,5-Diphenyl-2,4-pentadien-1-one 98+%
1,5-Diphenyl-2,4-pentadien-1-one

Purity: 98+% CAS No.: 614-57-3 Molecular Formula: C17H14O Formula Weight: 234.30 Melting Point: 100-102° Beilstein Reference: 1870052

Formula: C₁₇H₁₄O
MW: 234.30 g/mol
Melting Pt: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00020692
CAS Number: 614-57-3
EINECS: 210-385-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR