4-(2-Chloro-6-fluorophenyl)but-3-en-2-one 98%
Supplier: Thermo Scientific Chemicals
A15083-03
A15083-06
CAAAAA15083-03EA
41.87
CAD
CAAAAA15083-03
CAAAA15083-06
CAAAA15083-14
4-(2-Chloro-6-fluorophenyl)but-3-en-2-one 98%
4-(2-Chloro-6-fluorophenyl)but-3-en-2-one
Purity: 98%
CAS No.: 175136-82-0
Molecular Formula: C10H8ClFO
Formula Weight: 198.63
Refractive Index: 1.5800
Boiling Point: 115°/0.8mm
MDL No.: MFCD00051608
Formula:
C₁₀H₈ClFO Boiling Pt: 115 °C/0.8 mm Storage Temperature: Ambient |
MDL Number:
MFCD00051608 CAS Number: 175136-82-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...