3-(4-Carboxyphenyl)propionic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15081-09
A15081-22
A15081-06
A15081-14
CAAAAA15081-09EA
265.44
CAD
CAAAAA15081-09
CAAAAA15081-22
CAAAA15081-06
CAAAA15081-14
3-(4-Carboxyphenyl)propionic acid 98%
3-(4-Carboxyphenyl)propionic acid
Purity: 98%
CAS No.: 38628-51-2
Molecular Formula: C10H10O4
Formula Weight: 194.19
Melting Point: 288-291°
Beilstein Reference: 3271796
MDL No.: MFCD00016560
Formula:
C₁₀H₁₀O₄ Melting Pt: 288…291 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016560 CAS Number: 38628-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...