Sodium 2-Bromoethanesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15076-14
A15076-22
A15076-36
CAAAA15076-14EA
61.78
CAD
CAAAA15076-14
CAAAA15076-22
CAAAA15076-36
Sodium 2-Bromoethanesulfonate 98%
Sodium 2-Bromoethanesulfonate
Purity: 98%
CAS No.: 4263-52-9
Molecular Formula: BrCH2CH2SO3Na
Formula Weight: 211.02
Melting Point: ca 284° dec.
Beilstein Reference: 4590828
MDL No.: MFCD00007530
Formula:
C₂H₅BrO₃S Melting Pt: ca 284 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00007530 CAS Number: 4263-52-9 EINECS: 224-244-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...