3,5-Di-tert-butyl-4-hydroxyphenylacetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15015-03
A15015-06
CAAAA15015-03EA
84.53
CAD
CAAAA15015-03
CAAAA15015-06
3,5-Di-tert-butyl-4-hydroxyphenylacetic acid 98%
3,5-Di-tert-butyl-4-hydroxyphenylacetic acid
Purity: 98%
CAS No.: 1611-03-6
Molecular Formula: C16H24O3
Formula Weight: 264.37
Melting Point: 154-158°
MDL No.: MFCD00156138
Formula:
C₁₆H₂₄O₃ MW: 264.37 g/mol Melting Pt: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00156138 CAS Number: 1611-03-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...