p-Xylene-2-sulfonic acid hydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15007-14
A15007-22
A15007-36
CAAAA15007-14EA
79
CAD
CAAAA15007-14
CAAAA15007-22
CAAAA15007-36
p-Xylene-2-sulfonic acid hydrate 99%
p-Xylene-2-sulfonic acid hydrate
Purity: 99%
CAS No.: 609-54-1
Molecular Formula: C8H10O3S.xH2O
Formula Weight: 186.23(anhy)
Melting Point: 84-86°
MDL No.: MFCD00151010
Formula:
C₈H₁₀O₃S Melting Pt: 84…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151010 CAS Number: 609-54-1 EINECS: 210-193-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...