4'-Cyanoacetanilide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14941-06
A14941-14
CAAAA14941-06EA
47.4
CAD
CAAAA14941-06
CAAAA14941-14
4'-Cyanoacetanilide 98%
4'-Cyanoacetanilide
Purity: 98%
CAS No.: 35704-19-9
Molecular Formula: C9H8N2O
Formula Weight: 160.18
Melting Point: 204-206°
MDL No.: MFCD00001814
Formula:
C₉H₈N₂O Melting Pt: 204…206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001814 CAS Number: 35704-19-9 EINECS: 252-685-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...