4-Chromanol 97%

Supplier: Thermo Scientific Chemicals

A14940-03 A14940-06
CAAAA14940-03EA 66.52 CAD
CAAAA14940-03 CAAAA14940-06 CAAAA14940-14
4-Chromanol 97%
4-Chromanol

Purity: 97% CAS No.: 1481-93-2 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 110°(230°F) Melting Point: 38-42° Boiling Point: 79-81°/5mm Beilstein Reference: 130800 MDL No.: MFCD00006849

Formula: C₉H₁₀O₂
Boiling Pt: 79…81 °C/5 mm
Melting Pt: 38…42 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006849
CAS Number: 1481-93-2
EINECS: 216-041-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR