Potassium tetraoxalate dihydrate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Oxalic acid, potassium salt dihydrate, Potassium trihydrogen dioxalate dihydrate
A14933-0B
A14933-0I
A14933-30
CAAAA14933-0BEA
214.88
CAD
CAAAA14933-0B
CAAAA14933-0I
CAAAA14933-30
Potassium tetraoxalate dihydrate 98+%
Potassium tetraoxalate dihydrate
Applications: Metal polishing determination of pH
Purity: 98+%
CAS No.: 6100-20-5
Molecular Formula: KH3(C2O4)2.2H2O
Formula Weight: 254.19 (218.16anhy)
Merck Reference: 14,7689
UN No.: 3288
MDL No.: MFCD00150443
Formula:
C₄H₃KO₈·2H₂O MW: 254.19 g/mol (218.16 anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150443 CAS Number: 6100-20-5 EINECS: 204-874-6 Merck Index: 14,7689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...