Trimethylamine-N-oxide dihydrate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trimethylamine oxide dihydrate

A14916-14 A14916-22 A14916-36
CAAAA14916-14EA 104.75 CAD
CAAAA14916-14 CAAAA14916-22 CAAAA14916-36
Trimethylamine-N-oxide dihydrate 98+%
Trimethylamine N-oxide dihydrate

Fieser: 1,1230 6,624 7,392 9,488 10,423 12,533 13,325 14,329 16,360


Purity: 98+% CAS No.: 62637-93-8 Molecular Formula: (CH3)3NO.2H2O Formula Weight: 111.14 (75.11anhy) Melting Point: 96-101° Beilstein Reference: 3612927 Merck Reference: 14,9710 MDL No.: MFCD00149077

Formula: C₃H₉NO·2H₂O
MW: 111.14 g/mol (75.11 anhy)
Melting Pt: 96…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00149077
CAS Number: 62637-93-8
EINECS: 214-675-6
Merck Index: 14,9710

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR