Fluorene-2-carbaldehyde 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Fluorene-2-carboxaldehyde

A14911-06 A14911-14 A14911-22
CAAAA14911-06EA 124.5 CAD
CAAAA14911-06 CAAAA14911-14 CAAAA14911-22
Fluorene-2-carbaldehyde 99%
Fluorene-2-carbaldehyde

Purity: 99% CAS No.: 30084-90-3 Molecular Formula: C14H10O Formula Weight: 194.23 Melting Point: 82-86° Beilstein Reference: 2255852

Formula: C₁₄H₁₀O
Melting Pt: 82…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00001123
CAS Number: 30084-90-3
EINECS: 250-035-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR