4-Chloromethyl-2-methylthiazole hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14902-14
A14902-06
CAAAAA14902-14EA
984.34
CAD
CAAAAA14902-14
CAAAA14902-03
CAAAA14902-06
4-Chloromethyl-2-methylthiazole hydrochloride 98%
4-Chloromethyl-2-methylthiazole hydrochloride
Purity: 98%
CAS No.: 77470-53-2
Molecular Formula: C5H6ClNS.HCl
Formula Weight: 184.09
Storage Temperature: 2°C to 8°C
Melting Point: ca 165° dec.
Beilstein Reference: 108603
UN No.: 3261
MDL No.: MFCD00067725
Formula:
C₅H₇Cl₂NS Melting Pt: ∼ 165 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00067725 CAS Number: 77470-53-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...