(S)-Methyl-2-amino-4-methylpentanoate hydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Leu-OMe·HCl, H-Leu-OMe.HCl, L-Leucine methyl ester hydrochloride

A14897-14 A14897-22
CAAAA14897-14EA 91.64 CAD
CAAAA14897-14 CAAAA14897-22 CAAAA14897-36
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride 99%
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride

Purity: 99% CAS No.: 7517-19-3 Molecular Formula: C7H15NO2.HCl Formula Weight: 181.66 Melting Point: 149-151° Beilstein Reference: 3595133 Optical Rotation: +13° (c=2 in water) MDL No.: MFCD00012494

Formula: C7H16ClNO2
Melting Pt: 149…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00012494
CAS Number: 7517-19-3
EINECS: 231-375-0

Order Now

Specification Test Results

Optical rotation +13° (c=2 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR