Triphenylmethyl mercaptan 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Triphenylmethanethiol

A14888-06 A14888-14 A14888-22
CAAAA14888-06EA 61.62 CAD
CAAAA14888-06 CAAAA14888-14 CAAAA14888-22
Triphenylmethyl mercaptan 98+%
Triphenylmethyl mercaptan

Purity: 98+% CAS No.: 3695-77-0 Molecular Formula: (C6H5)3CSH Formula Weight: 276.40 Odor: Stench Melting Point: 103-108° Beilstein Reference: 2113168 MDL No.: MFCD00004854

Formula: C₁₉H₁₆S
MW: 276.40 g/mol
Melting Pt: 103…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00004854
CAS Number: 3695-77-0
EINECS: 223-020-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR