trans-2-Chloro-β-nitrostyrene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14882-06
A14882-14
CAAAA14882-06EA
114.23
CAD
CAAAA14882-06
CAAAA14882-14
CAAAA14882-22
trans-2-Chloro-β-nitrostyrene 98%
trans-2-Chloro-β-nitrostyrene
Purity: 98%
CAS No.: 3156-34-1
Molecular Formula: C8H6ClNO2
Formula Weight: 183.60
Melting Point: 46-49°
Beilstein Reference: 2328095
Formula:
C₈H₆ClNO₂ Melting Pt: 46…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00024820 CAS Number: 3156-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...