Methyl-3,5-di-tert-butyl-4-hydroxybenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14837-14
A14837-22
CAAAA14837-14EA
222.78
CAD
CAAAA14837-14
CAAAA14837-22
Methyl-3,5-di-tert-butyl-4-hydroxybenzoate 98%
Methyl-3,5-di-tert-butyl-4-hydroxybenzoate
Purity: 98%
CAS No.: 2511-22-0
Molecular Formula: C16H24O3
Formula Weight: 264.37
Melting Point: 164-166°
Beilstein Reference: 2218125
MDL No.: MFCD00017253
Formula:
C₁₆H₂₄O₃ MW: 264.37 g/mol Melting Pt: 164…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017253 CAS Number: 2511-22-0 EINECS: 219-729-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...