5-Acetylvaleric acid 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
6-Oxoheptanoic acid
A14810-06
A14810-09
A14810-18
CAAAAA14810-06EA
121.98
CAD
CAAAAA14810-06
CAAAA14810-09
CAAAA14810-18
5-Acetylvaleric acid 96%
5-Acetylvaleric acid
Soluble in water
Purity: 96%
CAS No.: 3128-07-2
Molecular Formula: C7H12O3
Formula Weight: 144.17
Refractive Index: 1.4490
Melting Point: 34-36°
Boiling Point: 138-140°/1.5mm
Solubility: Soluble in water
Beilstein Reference: 1362931
UN No.: 3261
MDL No.: MFCD00051479
Formula:
C₇H₁₂O₃ Boiling Pt: 138…140 °C/1.5 mm Melting Pt: 34…36 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051479 CAS Number: 3128-07-2 EINECS: 221-512-2 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...