Sodium 4-Hydroxybenzenesulfonate dihydrate 97%

Supplier: Thermo Scientific Chemicals
Warning

A14808-0E A14808-22 A14808-36
CAAAAA14808-0EEA 379.2 CAD
CAAAAA14808-0E CAAAA14808-22 CAAAA14808-36
Sodium 4-Hydroxybenzenesulfonate dihydrate 97%
Sodium-4-hydroxybenzenesulphonate dihydrate

Purity: 97% CAS No.: 10580-19-5 Molecular Formula: C6H5NaO4S.2H2O Formula Weight: 232.20 (196.17anhy) Odor: Odorless Melting Point: 300° Beilstein Reference: 4040891 MDL No.: MFCD00044734

Formula: C₆H₆O₄S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00044734
CAS Number: 10580-19-5
EINECS: 212-550-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR