cis-3-Chloroacrylic acid ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cis-3-chloroacrylicacid
A14798-03
A14798-06
CAAAA14798-03EA
76.79
CAD
CAAAA14798-03
CAAAA14798-06
CAAAA14798-14
cis-3-Chloroacrylic acid ≥98%
cis-3-Chloroacrylic acid
Purity: 98+%
CAS No.: 1609-93-4
Molecular Formula: C3H3ClO2
Formula Weight: 106.51
Storage Temperature: 2°C to 8°C
Melting Point: 60-62°
Beilstein Reference: 1720278
UN No.: 3261
MDL No.: MFCD00004368
Formula:
C₃H₃ClO₂ Melting Pt: 60…62 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004368 CAS Number: 1609-93-4 EINECS: 216-545-4 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...