Monomethyl phthalate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Methoxycarbonyl)benzoic acid, Methyl hydrogen phthalate
A14780-09
A14780-18
A14780-30
CAAAA14780-09EA
91.8
CAD
CAAAA14780-09
CAAAA14780-18
CAAAA14780-30
Monomethyl phthalate 97%
Monomethyl phthalate
Purity: 97%
CAS No.: 4376-18-5
Molecular Formula: C9H8O4
Formula Weight: 180.16
Melting Point: 81-84°
Beilstein Reference: 2048766
Formula:
C₉H₈O₄ MW: 180.16 g/mol Melting Pt: 81…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002466 CAS Number: 4376-18-5 EINECS: 224-476-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...