O-Methylisourea hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14773-30
A14773-09
A14773-18
CAAAAA14773-30EA
2529.58
CAD
CAAAAA14773-30
CAAAA14773-09
CAAAA14773-18
O-Methylisourea hydrochloride 98%
O-Methylisourea hydrochloride
Purity: 98%
CAS No.: 5329-33-9
Molecular Formula: C2H6N2O.HCl
Formula Weight: 110.54
Melting Point: ca 116° dec.
Beilstein Reference: 3909619
MDL No.: MFCD00035043
Formula:
C₂H₇ClN₂O Melting Pt: ca 116 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00035043 CAS Number: 5329-33-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...