Acid red 52

Supplier: Thermo Scientific Chemicals

A14769-06 A14769-14
CAAAA14769-06EA 83.42 CAD
CAAAA14769-06 CAAAA14769-14
Acid red 52
Acid red 52

Fluorescent dye which uses laser-induced fluorescence for quantification of cellular proteins


CAS No.: 3520-42-1 Molecular Formula: C27H29N2NaO7S2 Formula Weight: 580.66 Beilstein Reference: 3886008 MDL No.: MFCD00010180

Formula: C₂₇H₃₀N₂O₇S₂
Storage Temperature: Ambient
MDL Number: MFCD00010180
CAS Number: 3520-42-1
EINECS: 222-529-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR