trans-3-(3-Pyridyl)acrylic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14747-06
A14747-14
A14747-30
CAAAAA14747-06EA
54.83
CAD
CAAAAA14747-06
CAAAAA14747-14
CAAAA14747-30
trans-3-(3-Pyridyl)acrylic acid 99%
trans-3-(3-Pyridyl)acrylic acid
Purity: 99%
CAS No.: 19337-97-4
Molecular Formula: C8H7NO2
Formula Weight: 149.15
Melting Point: 232-235°
Beilstein Reference: 113290
MDL No.: MFCD00006410
Formula:
C₈H₇NO₂ Melting Pt: 232…235 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006410 CAS Number: 19337-97-4 EINECS: 214-424-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...