4-Decyloxybenzoic acid 96%

Supplier: Thermo Scientific Chemicals

A14703-06 A14703-14 A14703-22
CAAAA14703-06EA 73.63 CAD
CAAAA14703-06 CAAAA14703-14 CAAAA14703-22
4-Decyloxybenzoic acid 96%
4-Decyloxybenzoic acid

Applications: synthesis of Liquid crystals
m.p. 97°(S), 122°(N), 142°(I)


Purity: 96% CAS No.: 5519-23-3 Molecular Formula: C17H26O3 Formula Weight: 278.39 Melting Point: 97°(S) Beilstein Reference: 2113730

Formula: C₁₇H₂₆O₃
Melting Pt: 97 °C(S)
Storage Temperature: Ambient
MDL Number: MFCD00020360
CAS Number: 5519-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR