4-(p-Tolyl)butyric acid 97%
Supplier: Thermo Scientific Chemicals
A14680-03
A14680-06
A14680-14
CAAAA14680-03EA
85.79
CAD
CAAAA14680-03
CAAAA14680-06
CAAAA14680-14
4-(p-Tolyl)butyric acid 97%
4-(p-Tolyl)butyric acid
Purity: 97%
CAS No.: 4521-22-6
Molecular Formula: C11H14O2
Formula Weight: 178.23
Melting Point: 55-59°
Beilstein Reference: 639705
MDL No.: MFCD00021786
Formula:
C₁₁H₁₄O₂ Melting Pt: 55…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021786 CAS Number: 4521-22-6 EINECS: 224-848-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...