3-Octen-2-one 97%
Supplier: Thermo Scientific Chemicals
A14652-09
A14652-18
A14652-30
CAAAA14652-09EA
73.15
CAD
CAAAA14652-09
CAAAA14652-18
CAAAA14652-30
3-Octen-2-one 97%
3-Octen-2-one
Purity: 97%
CAS No.: 1669-44-9
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 54°(129°F)
Refractive Index: 1.4480
Odor: Pleasant
Boiling Point: 60°/16mm
Beilstein Reference: 1742230
UN No.: 1224
MDL No.: MFCD00015565
Formula:
C₈H₁₄O MW: 126.20 g/mol |
MDL Number:
MFCD00015565 CAS Number: 1669-44-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...