3-Bromochromone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14591-03
A14591-06
CAAAA14591-01EA
43.72
CAD
CAAAA14591-01
CAAAA14591-03
CAAAA14591-06
3-Bromochromone 97%
3-Bromochromone
Purity: 97%
CAS No.: 49619-82-1
Molecular Formula: C9H5BrO2
Formula Weight: 225.05
Melting Point: 94-98°
Beilstein Reference: 125684
Formula:
C₉H₅BrO₂ Melting Pt: 94…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017337 CAS Number: 49619-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...