Tetraethyl ((dimethylamino)methylene)bisphosphonate 97%

Supplier: Thermo Scientific Chemicals
Warning

A14581-06 A14581-14
CAAAA14581-06EA 82.63 CAD
CAAAA14581-06 CAAAA14581-14 CAAAA14581-22
Tetraethyl ((dimethylamino)methylene)bisphosphonate 97%
Tetraethyl ((dimethylamino)methylene)bisphosphonate

Fieser: 3,277 11,502


Purity: 97% CAS No.: 18855-52-2 Molecular Formula: C11H27NO6P2 Formula Weight: 331.29 Storage Temperature: 2°C to 8°C Refractive Index: 1.4510 Boiling Point: 138-140°/1mm Beilstein Reference: 1914890 MDL No.: MFCD00015128

Formula: C₁₁H₂₇NO₆P₂
Boiling Pt: 138…140 °C/1 mm
Storage Temperature: Refrigerator
MDL Number: MFCD00015128
CAS Number: 18855-52-2
EINECS: 242-627-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR