D-(+)-Cellobiose 98+%
Supplier: Thermo Scientific Chemicals
A14553-06
A14553-14
A14553-22
CAAAA14553-06EA
48.35
CAD
CAAAA14553-06
CAAAA14553-14
CAAAA14553-22
D-(+)-Cellobiose 98+%
D(+)-Cellobiose
Purity: 98+%
CAS No.: 528-50-7
Molecular Formula: C12H22O11
Formula Weight: 342.30
Physical Form: Powder
Appearance: White
Melting Point: 234° dec.
Beilstein Reference: 93795
Merck Reference: 14,1961
Optical Rotation: +34° (c=10 in water, 15h)
MDL No.: MFCD00136034
Formula:
C₁₂H₂₂O₁₁ Melting Pt: 234 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00136034 CAS Number: 528-50-7 EINECS: 208-436-5 Merck Index: 14,1961 |
Specification Test Results
Optical rotation | +34° (c=10 in water, 15h) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...