Tetraethyl methylenebisphosphonate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetraethyl methylenediphosphonate, Tetraethyl methylenebis(phosphonate)

A14532-06 A14532-14 A14532-22
CAAAA14532-06EA 138.41 CAD
CAAAA14532-06 CAAAA14532-14 CAAAA14532-22
Tetraethyl methylenebisphosphonate 98+%
Tetraethyl methylenebisphosphonate

Purity: 98+% CAS No.: 1660-94-2 Molecular Formula: C9H22O6P2 Formula Weight: 288.22 Flash Point: 110°(230°F) Refractive Index: 1.4400 Boiling Point: 151-153°/1.2mm Beilstein Reference: 1813241 MDL No.: MFCD00039887

Formula: C₉H₂₂O₆P₂
Boiling Pt: 151…153 °C (1,2 mmHg)
Density: 1,162
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039887
CAS Number: 1660-94-2
EINECS: 216-764-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR