Lawesson's reagent 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,4-Bis(4-methoxyphenyl)-2,4-dithioxo-1,3,2,4- dithiadiphosphetane, 2,4-Bis(4-methoxyphenyl)-1,3-dithia-2,4-diphosphetane 2,4-Disulfide, 2,4-Bis(4-methoxyphenyl)-1,3-dithia-2,4-diphosphetane 2,4-Disulphide

A14530-06 A14530-14 A14530-22 A14530-36
CAAAAA14530-06EA 33.65 CAD
CAAAAA14530-06 CAAAA14530-14 CAAAA14530-22 CAAAA14530-36
Lawesson's reagent 97%
Lawesson's reagent

Fieser: 8,327 9,49 10,39 11,54 12,59 13,38 15,37 16,37


Purity: 97% CAS No.: 19172-47-5 Molecular Formula: C14H14O2P2S4 Formula Weight: 404.47 Melting Point: 221-225° Beilstein Reference: 1024888 UN No.: 3134 MDL No.: MFCD00005171

Formula: C₁₄H₁₄O₂P₂S₄
MW: 404.45 g/mol
Melting Pt: 221…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00005171
CAS Number: 19172-47-5
EINECS: 242-855-4
UN: 3134
ADR: 4.3,III
Merck Index: 14,5391

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR