5,7-Dihydroxy-4-methyl-2H-chromen-2-one 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5,7-Dihydroxy-4-methylcoumarin

A14528-03 A14528-06
CAAAA14528-03EA 164.32 CAD
CAAAA14528-03 CAAAA14528-06
5,7-Dihydroxy-4-methyl-2H-chromen-2-one 98%
5,7-Dihydroxy-4-methyl-2H-chromen-2-one

Purity: 98% CAS No.: 2107-76-8 Molecular Formula: C10H8O4 Formula Weight: 192.17 Melting Point: 298-300° Beilstein Reference: 179133 MDL No.: MFCD00016966

Formula: C₁₀H₈O₄
MW: 192.17 g/mol
Melting Pt: 298…300 °C
Storage Temperature: Ambient
MDL Number: MFCD00016966
CAS Number: 2107-76-8
EINECS: 218-289-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR