2-Furoylacetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

A14497-03 A14497-06 A14497-14
CAAAA14497-03EA 51.82 CAD
CAAAA14497-03 CAAAA14497-06 CAAAA14497-14
2-Furoylacetonitrile 97%
2-Furoylacetonitrile
Formula: C₇H₅NO₂
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00052210
CAS Number: 31909-58-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR