Benzyloxyacetaldehyde diethyl acetal 98%
Supplier: Thermo Scientific Chemicals
Warning
|
A14474-09
A14474-18
A14474-30
CAAAA14474-09EA
147.57
CAD
CAAAA14474-09
CAAAA14474-18
CAAAA14474-30
Benzyloxyacetaldehyde diethyl acetal 98%
Benzyloxyacetaldehyde diethyl acetal
Purity: 98%
CAS No.: 42783-78-8
Molecular Formula: C6H5CH2OCH2CH(OCH2CH3)2
Formula Weight: 224.30
Flash Point: 79°(174°F)
Refractive Index: 1.4790
Boiling Point: 99-100°/0.6mm
Beilstein Reference: 3278795
MDL No.: MFCD00051786
Formula:
C₁₃H₂₀O₃ Boiling Pt: 99…100 °C/0.6 mm Density: 0.987 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051786 CAS Number: 42783-78-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...