6-Methyl-3(2H)-pyridazinone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14471-14
A14471-06
CAAAAA14471-14EA
718.9
CAD
CAAAAA14471-14
CAAAA14471-03
CAAAA14471-06
6-Methyl-3(2H)-pyridazinone 98%
6-Methyl-3(2H)-pyridazinone
Purity: 98%
CAS No.: 13327-27-0
Molecular Formula: C5H6N2O
Formula Weight: 110.12
Melting Point: 143-145°
Beilstein Reference: 109928
Formula:
C₅H₆N₂O Melting Pt: 143…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039720 CAS Number: 13327-27-0 EINECS: 236-367-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...