1-Naphthoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A14460-09 A14460-18 A14460-30
CAAAA14460-09EA 78.37 CAD
CAAAA14460-09 CAAAA14460-18 CAAAA14460-30
1-Naphthoyl chloride 98%
1-Naphthoyl chloride

Purity: 98% CAS No.: 879-18-5 Molecular Formula: C11H7ClO Formula Weight: 190.63 Flash Point: 110°(230°F) Refractive Index: 1.6525 Odor: Pungent Melting Point: 16-19° Boiling Point: 118°/0.1mm Beilstein Reference: 775785 UN No.: 3265 MDL No.: MFCD00004002

Formula: C₁₁H₇ClO
MW: 190.63 g/mol
Boiling Pt: 118 °C (0.1 mmHg)
Melting Pt: 16…19 °C
Density: 1.265
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004002
CAS Number: 879-18-5
EINECS: 212-903-9
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR